missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR5M9 Polyclonal Antibody
GREENER_CHOICE

Codice prodotto. 16668684 Sfoglia Tutto Thermo Scientific Prodotti
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
16668684 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Questo articolo non è restituibile. Consulta la politica di reso
Codice prodotto. 16668684 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PA5102212

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR5M9.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifica

Antigen OR5M9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR5M9
Gene Accession No. Q8NGP3
Gene Alias Olfactory receptor 5M9; olfactory receptor family 5 subfamily M member 9; olfactory receptor OR11-190; olfactory receptor, family 5, subfamily M, member 9; OR11-190; OR5M9
Gene Symbols OR5M9
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR5M9(Accession Q8NGP3), corresponding to amino acid residues N204-I254.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390162
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.