missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ORAOV1 Polyclonal Antibody

Product Code. 15624955
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15624955 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15624955 Supplier Invitrogen™ Supplier No. PA551030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ORAOV1 Polyclonal Antibody detects endogenous levels of ORAOV1 protein.

ORAOV1 is a protein coding gene. The complex LTO1:YAE1 functions as a target specific adapter that probably recruits apo-ABCE1 to the cytosolic iron-sulfur protein assembly (CIA) complex machinery. May be required for biogenesis of the large ribosomal subunit and initiation of translation. May play a role in the regulation of proline metabolism and ROS production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORAOV1
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Gene ORAOV1
Gene Accession No. Q8WV07
Gene Alias 2210010N10Rik; Lto1; oral cancer overexpressed 1; oral cancer overexpressed protein 1-A; oral cancer-overexpressed protein 1; Oral cancer-overexpressed protein 1 homolog; Oraov1; Protein LTO1 homolog; TAOS1; Tumor-amplified and overexpressed sequence 1
Gene Symbols ORAOV1
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human ORAOV1 (aa 81-130).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220064
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.