missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSR2 Polyclonal Antibody

Description
Immunogen sequence: THRSQELRGAA ATEGFLYVLL SHWVFVGAPR PPASDSWKKG LVPSAPPASR KMGSKALPAP IPLHPSLQL Highest antigen sequence identity to the following orthologs - mouse 31%, rat 31%.
OSR2 is a mammalian homolog of the Drosophila odd-skipped family of transcription factors.
Specifications
Specifications
| Antigen | OSR2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | OSR2 |
| Gene Accession No. | Q8N2R0 |
| Gene Alias | 5430409I15Rik; odd-skipped related 2; odd-skipped related transciption factor 2; Osr2; Osr2A; Osr2B; Protein odd-skipped-related 2 |
| Gene Symbols | OSR2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?