missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otx2 Antibody (1H12C4B5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37597-0.025ml
This item is not returnable.
View return policy
Description
Otx2 Monoclonal specifically detects Otx2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Otx2 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| OTX2 | |
| Purified recombinant fragment of human OTX2 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1H12C4B5 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Immunocytochemistry 1:200 | |
| homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2 | |
| Mouse | |
| 32 kDa | |
| 0.025 mL | |
| Core ESC Like Genes, Stem Cell Markers, Vision | |
| 5015 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction