missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otx2 Antibody (1H12C4B5), Novus Biologicals™
Mouse Monoclonal Antibody
198.00€ - 572.00€
Specifications
| Antigen | Otx2 |
|---|---|
| Clone | 1H12C4B5 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Immunocytochemistry 1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18775713
|
Novus Biologicals
NBP2-37597 |
0.1 mL |
572.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685785
|
Novus Biologicals
NBP2-37597-0.025ml |
0.025 mL |
198.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Otx2 Monoclonal specifically detects Otx2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Otx2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Immunocytochemistry 1:200 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2 | |
| OTX2 | |
| IgG1 | |
| Unpurified |
| 1H12C4B5 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Core ESC Like Genes, Stem Cell Markers, Vision | |
| Ascites with 0.03% Sodium Azide | |
| 5015 | |
| Purified recombinant fragment of human OTX2 expressed in E. Coli. | |
| Primary | |
| 32 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title