missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), Brilliant Violet™ 711, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 407-5743-80
This item is not returnable.
View return policy
Description
OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal antibody specifically detects OVA257-264 (SIINFEKL) peptide bound to H-2Kb in Mouse samples. It is validated for Flow Cytometry
Specifications
| OVA257-264 (SIINFEKL) peptide bound to H-2Kb | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| 25 μg | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| eBio25-D1.16 (25-D1.16) | |
| Brilliant Violet 711 | |
| H-2Kb-SIINFEKL; OVA257-264; OVA257-264 H-2Kb; OVA-Kb | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction