missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), Brilliant Violet™ 711, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
188.00€ - 385.00€
Specifications
| Antigen | OVA257-264 (SIINFEKL) peptide bound to H-2Kb |
|---|---|
| Clone | eBio25-D1.16 (25-D1.16) |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17992101
![]() |
Invitrogen™
407-5743-82 |
100 μg |
385.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17942121
|
Invitrogen™
407-5743-80 |
25 μg |
188.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal antibody specifically detects OVA257-264 (SIINFEKL) peptide bound to H-2Kb in Mouse samples. It is validated for Flow CytometrySpecifications
| OVA257-264 (SIINFEKL) peptide bound to H-2Kb | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| IgG1 κ | |
| Affinity chromatography |
| eBio25-D1.16 (25-D1.16) | |
| Flow Cytometry | |
| Brilliant Violet 711 | |
| Mouse | |
| Mouse | |
| H-2Kb-SIINFEKL; OVA257-264; OVA257-264 H-2Kb; OVA-Kb | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title