missing translation for 'onlineSavingsMsg'
Learn More

Oxybenzone-(phenyl-13C6), TRC

Product Code. 30410502
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410502 1 mg 1mg
30411434 5 mg 5mg
30410077 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410502 Supplier Toronto Research Chemicals Supplier No. TRCO8673011MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxybenzone-(phenyl-13C6)
Purity Grade Notes HPLC
Molecular Formula C813C6H12O3
Quantity 1 mg
InChI Formula InChI=1S/C14H12O3/c1-17-11-7-8-12(13(15)9-11)14(16)10-5-3-2-4-6-10/h2-9,15H,1H3/i2+1,3+1,4+1,5+1,6+1,10+1
SMILES O=C(C1=C(O)C=C(C=C1)OC)[13C]2=[13CH][13CH]=[13CH][13CH]=[13CH]2
IUPAC Name (2-hydroxy-4-methoxyphenyl)(phenyl-13C6)methanone
Molecular Weight (g/mol) 234.2
Formula Weight 234.1
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.