Learn More
p-Tyramine, TRC
Product Code. 17927524
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17927524 5 g 5g
30133578 25 g 25g
30133569 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17927524 Supplier Toronto Research Chemicals Supplier No. TRCT8985005G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51-67-2
Molecular Formula C8 H11 N O
Molecular Weight (g/mol) 137.18
Synonym Tyramine, Ritodrine Hydrochloride Imp. A (BP), Phenol, 4-(2-aminoethyl)-, Phenol, p-(2-aminoethyl)- (8CI), 4-(2-Aminoethyl)phenol, 2-(4-Hydroxyphenyl)ethanamine, 2-(4-Hydroxyphenyl)ethylamine, 2-(4'-Hydroxyphenyl)ethylamine, 2-(p-Hydroxyphenyl)ethylamine, 4-Hydroxy-β-phenylethylamine, 4-Hydroxyphenethylamine, 4-Hydroxyphenylethylamine, Benzeneethanamine, 4-hydroxy-, NSC 249188, Systogene, Tocosine, Tyramin, Tyrosamine, Uteramine, p-(2-Aminoethyl)phenol, p-Hydroxy-β-phenylethylamine, p-Hydroxyphenethylamine, p-Hydroxyphenylethylamine, p-Tyramine, p-β-Aminoethylphenol, α-(4-Hydroxyphenyl)-β-aminoethane, β-(4-Hydroxyphenyl)ethylamine
IUPAC Name 4-(2-aminoethyl)phenol
SMILES NCCc1ccc(O)cc1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C8H11NO/c9-6-5-7-1-3-8(10)4-2-7/h1-4,10H,5-6,9H2
Formula Weight 137.0841
Percent Purity >95%
Chemical Name or Material Tyramine
Grade HPLC