missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P2Y4 Polyclonal Antibody

Description
PA1-20619 detects P2Y4 in Human samples.
Receptor for UTP and UDP coupled to G-proteins that activate a phosphatidylinositol-calcium second messenger system. Not activated by ATP or ADP.
Specifications
Specifications
| Antigen | P2Y4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | P2RY4 |
| Gene Accession No. | P51582 |
| Gene Alias | NRU; P2P; P2RY4; P2Y ATP receptor 4; P2Y purinoceptor 4; P2Y4; P2Y4 receptor; P2Y4R; P2YR4; purinergic receptor P2Y G-protein coupled 4; purinergic receptor P2Y, G-protein coupled 4; purinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y4; RP13-26D14.5; UNR; Uridine nucleotide receptor |
| Gene Symbols | P2RY4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?