Learn More
P62 SQSTM1 Rabbit anti-Human, Unconjugated, Clone: 1D17, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80294-1-RR-20UL
Description
The P62, SQSTM1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human P62, SQSTM1. This antibody recognizes human antigen. The P62, SQSTM1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| P62 SQSTM1 | |
| Monoclonal | |
| Unconjugated | |
| 20 μL | |
| 8878 | |
| IgG |
| Western Blot | |
| 1D17 | |
| Rabbit | |
| Primary | |
| Human |