Learn More
P62 SQSTM1 Rabbit anti-Human, Unconjugated, Clone: 1D17, Proteintech
Rabbit Recombinant Antibody
Brand:  Proteintech 80294-1-RR-20UL
 View more versions of this product
Product Code. 17442453
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
The P62, SQSTM1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human P62, SQSTM1. This antibody recognizes human antigen. The P62, SQSTM1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
P62 SQSTM1
Monoclonal
Unconjugated
20 μL
8878
IgG
Western Blot
1D17