Learn More
p63 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Product Code. 16850566 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16850566 100 μL 100µL
17781249 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16850566 Supplier Proteintech Supplier No. CL48812143100UL
Rabbit Polyclonal Antibody
The p63 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human p63. This antibody recognizes human, mouse, rat antigen. The p63 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Antigen p63
Applications Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Polyclonal
Concentration 1200 μg/mL
Conjugate CoraLite 488
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 8626
Target Species Human, Rat, Mouse
Isotype IgG