missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
p70 S6 Kinase beta/S6K2 Polyclonal specifically detects p70 S6 Kinase beta/S6K2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | p70 S6 Kinase beta/S6K2 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, KLS, p70 ribosomal S6 kinase beta, p70 S6 kinase beta, p70 S6KB, p70 S6K-beta, p70(S6K)-beta, p70-beta, P70-beta-1, P70-beta-2, p70-S6K 2, P70S6K2, p70S6Kb, ribosomal protein S6 kinase beta-2, ribosomal protein S6 kinase, 70kD, polypeptide 2,70 kDa ribosomal protein S6 kinase 2, ribosomal protein S6 kinase, 70kDa, polypeptide 2, S6K2, S6K-beta, S6K-beta2, S6K-beta-2, serine/threonine-protein kinase 14 beta, Serine/threonine-protein kinase 14B, SRK, STK14BS6 kinase-related kinase |
| Gene Symbols | RPS6KB2 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VDSPDDTALSESANQAFLGFTYVAPSVLDSIKEGFSFQPKLRSPRRLNSSPRAPVSPLKFSPFEGFRPSPSLPEPTELPLPPLLPPPPPSTTAPLPIRPPSGTKK |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?