missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC25A43 Polyclonal specifically detects SLC25A43 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | SLC25A43 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | solute carrier family 25 member 43, solute carrier family 25, member 43 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human slc25a43 (NP_660348.2). MATWRRDGRLTGGQRLLCAGLAGTLSLSLTAPLELATVLAQVGVVRGHARGPWATGHRVWRAEGLRALWKGNAVACLRLFPCSAVQLAAYRKFVVLFTDD |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?