missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan RhO/RAC/CDC42 Antibody (3L74), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-29740
This item is not returnable.
View return policy
Description
pan RhO/RAC/CDC42 Monoclonal specifically detects pan RhO/RAC/CDC42 in Human, Rat samples. It is validated for Western Blot.
Specifications
| pan RhO/RAC/CDC42 | |
| Monoclonal | |
| Unconjugated | |
| No buffer with No Preservative | |
| h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA | |
| Rabbit | |
| 22 kDa | |
| 0.1 mL | |
| mTOR Pathway, Signal Transduction, Wnt Signaling Pathway | |
| 387 | |
| Human, Rat | |
| IgG |
| Western Blot | |
| 3L74 | |
| Western Blot 1:100-1:2000 | |
| P61586 | |
| RHOA | |
| KLH-conjugated, synthetic peptide corresponding to amino acids 73-104 of human Rho (-A, -B, -C), with two terminal lysines added to increase peptide solubility. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Recognizes Rho (-A, -B, -C). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction