missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan RhO/RAC/CDC42 Antibody (3L74), Novus Biologicals™
Rabbit Monoclonal Antibody
191.00€ - 657.00€
Specifications
| Antigen | pan RhO/RAC/CDC42 |
|---|---|
| Clone | 3L74 |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18136802
|
Novus Biologicals
NBP2-29740-0.025ML |
0.025 mL |
191.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18120524
|
Novus Biologicals
NBP2-29740 |
0.1 mL |
657.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
pan RhO/RAC/CDC42 Monoclonal specifically detects pan RhO/RAC/CDC42 in Human, Rat samples. It is validated for Western Blot.Specifications
| pan RhO/RAC/CDC42 | |
| Western Blot 1:100-1:2000 | |
| Monoclonal | |
| Rabbit | |
| mTOR Pathway, Signal Transduction, Wnt Signaling Pathway | |
| No buffer with No Preservative | |
| h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA | |
| RHOA | |
| IgG | |
| Affinity Purified | |
| Recognizes Rho (-A, -B, -C). |
| 3L74 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| P61586 | |
| 387 | |
| KLH-conjugated, synthetic peptide corresponding to amino acids 73-104 of human Rho (-A, -B, -C), with two terminal lysines added to increase peptide solubility. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 22 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title