Learn More
Paraffin, liquid, pure
Product Code. 10540614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10540614 2.5 L Glass bottle 2.5 L
10689014 250 mL Glass bottle 250mL
10361124 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10540614 Supplier Thermo Scientific Acros Supplier No. 171400025
  • ONLINE EXCLUSIVE PRICE
    110.00€
    94.65€ / 2.5 L
    Save 15.35€
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 8012-95-1 | MFCD00131611
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
Synonym delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
PubChem CID 68245
ChEBI CHEBI:38701
IUPAC Name 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
SMILES *
Specifications
Melting Point -24.0°C
Color Colorless
Density 0.8800g/mL
Boiling Point 300.0°C
Flash Point 218°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 2.5 L
Merck Index 14, 7186
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble
Viscosity 65-75 mm2/s (40°C)
Grade Pure
Physical Form Oily Liquid
Chemical Name or Material Paraffin, liquid, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Paraffin, liquid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.