missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAX4 Monoclonal Antibody (3C12)
GREENER_CHOICE

Product Code. 12817822
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12817822 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12817822 Supplier Invitrogen™ Supplier No. MA515798

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-15798 targets PAX4 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15798 immunogen is purified recombinant fragment of human PAX4 expressed in E. Coli. MA5-15798 detects PAX4 which has a predicted molecular weight of approximately 38kDa.

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The paired box 4 gene is involved in pancreatic islet development and mouse studies have demonstrated a role for this gene in differentiation of insulin-producing beta cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C12
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene PAX4
Gene Accession No. O43316
Gene Alias KPD; MGC129960; MODY9; paired box 4; paired box gene 4; paired box protein Pax-4; paired domain gene 4; Pax4; Pax-4
Gene Symbols PAX4
Host Species Mouse
Immunogen Purified recombinant fragment of human PAX4 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5078
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.