missing translation for 'onlineSavingsMsg'
Learn More

PC aa C38:0-D39 Major, TRC

Product Code. 30428119
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428119 10 mg 10mg
30416838 100 mg 100mg
30412199 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428119 Supplier Toronto Research Chemicals Supplier No. TRCP48863210MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material PC aa C38:0-D39
Molecular Formula C46 D39 H53 N O8 P
Quantity 10 mg
Synonym (R)-2-((Icosanoyl-d39)oxy)-3-(stearoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate
InChI Formula InChI=1S/C46H92NO8P/c1-6-8-10-12-14-16-18-20-22-23-25-27-29-31-33-35-37-39-46(49)55-44(43-54-56(50,51)53-41-40-47(3,4)5)42-52-45(48)38-36-34-32-30-28-26-24-21-19-17-15-13-11-9-7-2/h44H,6-43H2,1-5H3/t44-/m1/s1/i1D3,6D2,8D2,10D2,12D2,14D2,16D2,18D2,20D2,22D2,23D2,25D2,27D2,29D2,31D2,33D2,35D2,37D2,39D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O[C@H](COC(=O)CCCCCCCCCCCCCCCCC)COP(=O)([O-])OCC[N+](C)(C)C
IUPAC Name [(2R)-2-(2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,20,20,20-nonatriacontadeuterioicosanoyloxy)-3-octadecanoyloxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
Molecular Weight (g/mol) 857.439
Formula Weight 856.901
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.