missing translation for 'onlineSavingsMsg'
Learn More

PCNA Rabbit anti-Human, Clone: 144, Novus Biologicals™

Product Code. 18298537 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18298537 50 μg 50µL
18228657 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18298537 Supplier Novus Biologicals Supplier No. NBP29005850UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PCNA Monoclonal specifically detects PCNA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNA
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 144
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Formulation 0.2 um filtered solution in PBS
Gene Alias cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Host Species Rabbit
Immunogen Recombinant Human PCNA Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5111
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.