missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDE6D Polyclonal specifically detects PDE6D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | PDE6D |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | O43924 |
| Gene Alias | GMP-PDE delta, PDED, phosphodiesterase 6D, cGMP-specific, rod, delta, Protein p17, retinal rod rhodopsin-sensitive cGMP 3'-5'-cyclic phosphodiesterase subunitdelta |
| Gene Symbols | PDE6D |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?