missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDK2 Recombinant Rabbit Monoclonal Antibody (K01_1W51)
GREENER_CHOICE

Product Code. 30825396
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825396 100 μL
30825146 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825396 Supplier Invitrogen™ Supplier No. MA563397

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PDK2 Recombinant Monoclonal Antibody for Western Blot

PDK2 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDK2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1W51
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene PDK2
Gene Accession No. Q15119, Q64536, Q9JK42
Gene Alias [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 2, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 2, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 2, mitochondrial; pyruvate dehydrogenase kinase, isozyme 2; LOC524075 protein; PDH kinase 2; PDHK2; PDK P45; PDK2; PDKII; pyruvate dehydrogenase; pyruvate dehydrogenase 2; pyruvate dehydrogenase kinase 2; pyruvate dehydrogenase kinase 2 subunit p45 (PDK2); Pyruvate dehydrogenase kinase isoform 2; pyruvate dehydrogenase kinase, isoenzyme 2; pyruvate dehydrogenase kinase, isozyme 2; pyruvate dehydrogenase, lipoamide, kinase isozyme 2, mitochondrial
Gene Symbols PDK2
Host Species Rabbit
Immunogen A synthetic peptide of human PDK2.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18604, 5164, 81530
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.