Learn More
Pejvakin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-75015
This item is not returnable.
View return policy
Description
Pejvakin Polyclonal antibody specifically detects Pejvakin in Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).