missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pejvakin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
222.00€ - 561.00€
Specifications
| Antigen | Pejvakin |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18297764
|
Novus Biologicals
NB110-75015-0.025ML |
0.025 mL |
222.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18725173
|
Novus Biologicals
NB110-75015 |
561.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Pejvakin Polyclonal antibody specifically detects Pejvakin in Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
| Pejvakin | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Q0ZLH3 | |
| 494513 | |
| IgG | |
| Unpurified | |
| Human. The peptide is homologous in mouse. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Autosomal recessive deafness type 59 protein, deafness, autosomal recessive 59, pejvakin, PJVK | |
| DFNB59 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title