missing translation for 'onlineSavingsMsg'
Learn More

Penicillin Antibody (P2B9), Novus Biologicals™

Product Code. 18793084 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
0.03mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18793084 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18793084 Supplier Novus Biologicals Supplier No. NBP1785880.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Penicillin Monoclonal specifically detects Penicillin in Fungi samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Penicillin
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone P2B9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
Gene Alias Ampicillin, Benzylpenicillin, dicloxacillin, flucloxacillin, methicillin, PCN, Penicilin, Penicillin G, Phenoxymethylpenicillin
Host Species Mouse
Immunogen Penicillin-BSA
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Chemical.
Target Species Other
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.