missing translation for 'onlineSavingsMsg'
Learn More

Pentaethylene Glycol, TRC

Product Code. 30009977
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009977 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30009977 Supplier Toronto Research Chemicals Supplier No. TRCP2272301G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4792-15-8
Molecular Formula C10H22O6
Quantity 1 g
Synonym 3,6,9,12-Tetraoxatetradecane-1,14-diol, 1,2-Bis[2-(2-hydroxyethoxy)ethoxy]ethane, 2,4,6,8-Tetraoxadecane-1,10-diol, 3,4,7,8,11,12,15,16-Octaoxaoctadecane-1,18-diol
InChI Formula InChI=1S/C10H22O6/c11-1-3-13-5-7-15-9-10-16-8-6-14-4-2-12/h11-12H,1-10H2
SMILES OCCOCCOCCOCCOCCO
IUPAC Name 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol
Molecular Weight (g/mol) 238.28
Formula Weight 238.1416
Chemical Name or Material Pentaethylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.