Learn More
Pentafluorophenyl pyridine-2-carboxylate, 95%, Thermo Scientific™
Product Code. 10041714
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041714 250 mg Amber Glass Bottle 250mg
10777855 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10041714 Supplier Thermo Scientific Maybridge Supplier No. CC04026CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Chemical Identifiers
CAS 188837-53-8
Molecular Formula C12H4F5NO2
Molecular Weight (g/mol) 289.16
MDL Number MFCD02916457
InChI Key KFJSLIBBYMXLTG-UHFFFAOYSA-N
Synonym pentafluorophenyl pyridine-2-carboxylate, pentafluorophenyl picolinate, 2-pyridinecarboxylicacid, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl pyridine-2-carboxylate, picolinic acid pentafluorophenyl ester, 2-pentafluorophenoxy carbonyl pyridine, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-2-carboxylate, 2-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 2754830
IUPAC Name (2,3,4,5,6-pentafluorophenyl) pyridine-2-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=N2)C(F)=C1F
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 289.16
Percent Purity 95%
Chemical Name or Material Pentafluorophenyl pyridine-2-carboxylate