Learn More
Perilipin-2/ADFP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB110-40877SS
 View more versions of this product
Product Code. 18715203
  • ONLINE EXCLUSIVE PRICE
    315.00€
    297.15€ / 25µL
    Save 17.85€
    5% Off
Description
Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
PLIN2
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 35041644), Dual RNAscope ISH-IHC
Affinity Purified