missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin-2/ADFP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
297.15€ - 603.75€
Specifications
| Dilution | Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 35041644), Dual RNAscope ISH-IHC |
|---|---|
| Classification | Polyclonal |
| Gene Symbols | PLIN2 |
| Purification Method | Affinity Purified |
| Content And Storage | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18715203
|
Novus Biologicals
NB110-40877SS |
315.00€ 297.15€ / 25µL Save 17.85€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18797653
|
Novus Biologicals
NB110-40877 |
639.00€ 603.75€ / 0.10mL Save 35.25€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.Specifications
| Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 35041644), Dual RNAscope ISH-IHC | |
| PLIN2 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Polyclonal | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title