missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB110-40760
This item is not returnable.
View return policy
Description
Perilipin Polyclonal specifically detects Perilipin in Human, Mouse, Rat, Porcine, Fish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Imaging Mass Cytometry.
Specifications
| Perilipin | |
| Polyclonal | |
| Unconjugated | |
| Tris-Glycine and 0.15 M NaCl with 0.05% Sodium Azide | |
| PERI, perilipin, perilipin 1, perilipin-1, PLINLipid droplet-associated protein | |
| Rabbit | |
| 60 kDa | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
| 5346 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 2 ug/mL, Flow Cytometry 5 - 10 ug/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Imaging Mass Cytometry | |
| O60240 | |
| PLIN1 | |
| This Perilipin Antibody was developed against a synthetic peptide made to a region between residues 450-522 (C-terminus) of the human Perilipin protein. [Swiss-Prot# O60240] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Fish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction