missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
244.00€ - 518.00€
Specifications
| Antigen | Perilipin |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug/mL, Flow Cytometry 5 - 10 ug/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Imaging Mass Cytometry |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18731414
|
Novus Biologicals
NB110-40760SS |
0.025 mL |
244.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18770484
|
Novus Biologicals
NB110-40760 |
0.1 mL |
518.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Perilipin Polyclonal specifically detects Perilipin in Human, Mouse, Rat, Porcine, Fish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Imaging Mass Cytometry.Specifications
| Perilipin | |
| Western Blot 2 ug/mL, Flow Cytometry 5 - 10 ug/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Imaging Mass Cytometry | |
| Polyclonal | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
| Tris-Glycine and 0.15 M NaCl with 0.05% Sodium Azide | |
| PERI, perilipin, perilipin 1, perilipin-1, PLINLipid droplet-associated protein | |
| PLIN1 | |
| IgG | |
| Affinity Purified | |
| 60 kDa |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Fish | |
| O60240 | |
| 5346 | |
| This Perilipin Antibody was developed against a synthetic peptide made to a region between residues 450-522 (C-terminus) of the human Perilipin protein. [Swiss-Prot# O60240] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title