Product Code. 10294150
Shop All Fisher Chemical Products
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10294150
25 L
Metal Can
25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
10294150
Supplier
Fisher Chemical
Supplier No.
P/1400/25
Estimated Shipment: 15-10-2026Log in to see stock.
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Chemical Identifiers
CAS
64742-49-0
Molecular Formula
C6H14
Molecular Weight (g/mol)
86.178
MDL Number
MFCD00081849
InChI Key
AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID
7892
SMILES
CCCC(C)C
Specifications
Melting Point
-130°C
Quantity
25 L
Packaging
Metal Can
Physical Form
Liquid
Vapor Pressure
514mmHg 25
Boiling Point
30°C
IUPAC Name
2-methylpentane
Formula Weight
72.15g/mol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?