missing translation for 'onlineSavingsMsg'
Learn More

PEX5 Monoclonal Antibody (OTI6E9), TrueMAB™, OriGene

Product Code. 17554221
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17554221 30 μL 30µL
17534221 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17554221 Supplier Origene Technologies Supplier No. TA501430S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PEX5 Monoclonal antibody specifically detects PEX5 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen PEX5
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI6E9
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CORO1A
Gene Accession No. P31146
Gene Alias AW212715; ESTM1; LOW QUALITY PROTEIN: peroxisomal biogenesis factor 5; PBD2A; PBD2B; peroxin 5; peroxin-5; peroxisomal biogenesis factor 5; peroxisomal C-terminal targeting signal import receptor; peroxisomal targeting signal 1 (SKL type) receptor; peroxisomal targeting signal 1 receptor; peroxisomal targeting signal 1 receptor; peroxisomal biogenesis factor 5; peroxisomal targeting signal import receptor; peroxisomal targeting signal receptor 1; peroxisome biogenesis factor 5; peroxisome receptor 1; PEX5; Pex5_predicted; PTS1 BP; PTS1 receptor; PTS1-BP; PTS1R; Pxr1; PXR1P; RCDP5; X83306
Host Species Mouse
Immunogen Full length human recombinant protein of human CORO1A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11151
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.