missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 670462
GREENER_CHOICE

Product Code. 18094862
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18094862 10 mg 10mg
18054872 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18094862 Supplier Tocris Bioscience™ Supplier No. 3316/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective CK1ε and CK1δ inhibitor

Potent and selective casein kinase 1ε (CK1ε) and CK1δ inhibitor (IC50 values are 7.7 and 14 nM respectively) that displays > 30-fold selectivity over 42 other common kinases. Inhibits PER protein nuclear translocation (EC50 = 290 nM) causing phase shifts in circadian rhythms and attenuates methamphetamine-stimulated locomotion in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 950912-80-8
Molecular Formula C19H22Cl2FN5

Specifications

Quantity 10 mg
Formula Weight 410.32
Purity >99%
Molecular Weight (g/mol) 410.318
Chemical Name or Material PF 670462
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.