missing translation for 'onlineSavingsMsg'
Learn More

PGL2 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18611962 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18611962 0.02 mL 0.02mL
18693402 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18611962 Supplier Novus Biologicals Supplier No. NBP2940390.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PGL2 Polyclonal antibody specifically detects PGL2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGL2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias C11orf79, FLJ20487, hSDH5, paraganglioma or familial glomus tumors 2, PGL2chromosome 11 open reading frame 79, SDH5SDH assembly factor 2, succinate dehydrogenase assembly factor 2, mitochondrial, succinate dehydrogenase complex assembly factor 2, Succinate dehydrogenase subunit 5, mitochondrial
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-166 of human SDHAF2 (NP_060311.1). MAVSTVFSTSSLMLALSRHSLLSPLLSVTSFRRFYRGDSPTDSQKDMIEIPLPPWQERTDESIETKRARLLYESRKRGMLENCILLSLFAKEHLQHMTEKQLNLYDRLINEPSNDWDIYYWATEAKPAPEIFENEVMALLRDFAKNKNKEQRLRAPDLEYLFEKPR
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 54949
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.