missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PGM2 Polyclonal Antibody

Product Code. 15717111 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15717111 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15717111 Supplier Invitrogen™ Supplier No. PA559247

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PHDKGISQAI EENLEPWPQA WDDSLIDSSP LLHNPSASIN NDYFEDLKKY CFHRSVNRET KVKFVH Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Catalyzes the conversion of the nucleoside breakdown products ribose-1-phosphate and deoxyribose-1-phosphate to the corresponding 5-phosphopentoses. May also catalyze the interconversion of glucose-1-phosphate and glucose-6-phosphate. Has low glucose 1,6-bisphosphate synthase activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGM2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PGM2
Gene Accession No. Q96G03
Gene Alias Glucose phosphomutase 2; MSTP006; PGM 2; PGM2; Phosphodeoxyribomutase; phosphoglucomutase 2; phosphoglucomutase-2; Phosphopentomutase
Gene Symbols PGM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PGM2. Recombinant protein control fragment (Product #RP-96732).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55276
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.