missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH Sensor InLab™ Science Pro-DES™
GREENER_CHOICE

Product Code. 31454049 Shop All Mettler Toledo Products
Change view
Click to view available options
Connector Types:
Direct
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types
31454049 Direct
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31454049 Supplier Mettler Toledo™ Supplier No. 31027064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A glass body, combination pH electrode with integrated temperature sensor, DES sensor head, bridge electrolyte and movable glass sleeve junction. Designed for difficult applications like organic solvents, emulsions and protein containing samples.

  • Self-cleaning junction: The high electrolyte outflow prevents the pollutants from entering and clogging the junction, which would interfere with the reading.
  • Easy to clean: The movable sleeve-junction can be easily and thoroughly cleaned and allows for a simple refill of the reference electrolyte.
  • Always the right temperature: The integrated temperature sensor guarantees measurements are always performed with the correct temperature compensation.
  • DES™—Diagnosable Efficient Sustainable: Increased efficiency with sensor diagnostics: faster problem solving, less downtime, maximized sensor lifespan, and support of preventive maintenance.
  • Stay ahead: The sensor's diagnostic capability means peace of mind for the sensor condition. It supports preventive maintenance, maximizes the sensor's lifespan, and prevents premature discard.
  • Sustainable investment: DES™ digital sensors are a smart choice. Retaining the measurement electronics and only replacing the sensor shaft at the end of its life, or the detachable cable that can be used for all sensors, helps reduce material waste and thus, the ecological footprint.
  • Boost productivity and efficiency: With diagnosed errors, the user guidance with troubleshooting recommendations allows for faster problem-solving, thus reducing downtime. It also minimizes handling errors therefore cutting down on the need for specialized training
TRUSTED_SUSTAINABILITY

Specifications

Connector Types Direct
Junction Movable Glass Sleeve
Material Glass
pH Range 0 to 12
Type Combination pH Electrode (Liquid-filled)
Internal Reference ARGENTHAL™ with Ag+-trap
Membrane A41
Probe NTC 30 kΩ
Reference Electrolyte 3 mol/L KCl
Sample Compatibility Alcohols,Gasoline,Organic Solvents
Sample Size ≥20 mL
Temperature (Metric) Range 0°C to100°C
Parameter pH
Sensor Types Combined Electrode
Refillable The Movable Sleeve-Junction Can Be Easily and Thoroughly Cleaned and Allows for a Simple Refill of the Reference Electrolyte
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.