missing translation for 'onlineSavingsMsg'
Learn More

Pharacine, TRC

Product Code. 17947334
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17947334 5 mg 5mg
17949064 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17947334 Supplier Toronto Research Chemicals Supplier No. TRCP2945505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63440-93-7
Molecular Formula C24H24O8
Molecular Weight (g/mol) 440.44
Synonym 3,8,15,20-Tetraoxatricyclo[20.2.2.210,13]octacosa-10,12,22,24,25,27-hexaene-2,9,14,21-tetrone, 1,4-Butanediol-terephthaloyl chloride cyclic dimer, Pharacin, Pharacine, X-Guard VCA 1, Polybutylene Terephthalate Cyclic Dimer, PBT Cyclic Dimer
IUPAC Name 3,8,15,20-tetraoxatricyclo[20.2.2.210,13]octacosa-1(25),10,12,22(26),23,27-hexaene-2,9,14,21-tetrone
SMILES O=C1OCCCCOC(=O)c2ccc(cc2)C(=O)OCCCCOC(=O)c3ccc1cc3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H24O8/c25-21-17-5-7-19(8-6-17)23(27)31-15-3-4-16-32-24(28)20-11-9-18(10-12-20)22(26)30-14-2-1-13-29-21/h5-12H,1-4,13-16H2
Formula Weight 440.1471
Percent Purity >95%
Chemical Name or Material Pharacine
Grade HPLC
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.