missing translation for 'onlineSavingsMsg'
Learn More

Phenol-OD, CDN

Product Code. 30319293
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319293 1 g 1g
30315108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319293 Supplier CDN Supplier No. CDND10581G
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenol-OD
CAS 1003-66-3
Molecular Formula C6 D H5 O
Quantity 1 g
Synonym Phenol-d (6 CI, 7 CI, 8 CI, 9 CI, ACI), Phenol-d, Phenol-O-d, Phenol-O-D, Phenol-OD, (Hydroxy-d)benzene
InChI Formula InChI=1 S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7 H/i/hD
SMILES [2 H]Oc1ccccc1
IUPAC Name deuteriooxybenzene
Molecular Weight (g/mol) 95.117
Formula Weight 95.048 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.