Learn More
Invitrogen™ PHF19 Polyclonal Antibody

Description
Positive Samples: Mouse brain, Rat brain; Cellular Location: Nucleus Immunogen sequence: MENRALDPGT RDSYGATSHL PNKGALAKVK NNFKDLMSKL TEGQYVLCRW TDGLYYLGKI KRVSSSKQSC LVTFEDNSKY WVLWKDIQHA GVPGEEPKCN ICLGKTSGPL NEILICGKCG LGYHQQCHIP IAGSADQPLL TPWFCRRCIF ALAVRVSLPS SPVPASPASS SGADQRLPSQ SLSSKQKGHT WALETDSASA TVLGQDL.
Specifications
Specifications
| Antigen | PHF19 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.63 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | PHF19 |
| Gene Accession No. | Q5T6S3, Q9CXG9 |
| Gene Alias | 3110009G19Rik; 3321402G02Rik; DKFZP727G051; hPCL3; MTF2L1; Pcl3; PHD finger protein 19; phf13; Phf19; polycomb like 3; polycomb-like 3; polycomb-like protein 3; si:dkeyp-51b7.1; TDRD19B; tudor domain containing 19B |
| Gene Symbols | PHF19 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.