missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHF8 Recombinant Rabbit Monoclonal Antibody (2H9D4)
GREENER_CHOICE

Product Code. 17216873 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17216873 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17216873 Supplier Invitrogen™ Supplier No. MA537995

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, C2C12, Mouse thymus Immunogen sequence: TVSNSPASQR TPGKRPIKRP AYWRTESEEE EENASLDEQD SLGACFKDAE YIYPSLESDD DDPALKSRPK KKKNSDDAPW SPKARVTPTL PKQDRPVREG T.

PHD finger protein 8 is a jumonji domain containing protein. Like other members of the jumonji family, PHF8 may therefore play a role in histone demethylation. Mutations in PHF8 lead to Siderius type X-linked mental retardation (MRXSSD), a mild to borderline type of mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF8
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2H9D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHF8
Gene Accession No. Q80TJ7, Q9UPP1
Gene Alias 9830141C09Rik; histone lysine demethylase PHF8; JHDM1F; jumonji C domain-containing histone demethylase 1F; Kiaa1111; mKIAA1111; MRXSSD; PHD finger protein 8; PHF8; RGD1561065; ZNF422
Gene Symbols PHF8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 800-900 of human PHF8 (Q9UPP1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23133, 317425, 320595
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.