missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phl p 1 Chimeric Recombinant Human Monoclonal Antibody (Clone 25)

Description
This antibody binds to the Timothy grass allergen Phl p 1 and shows cotss reactivity with natural group 1 allergens from a variety of grass species such as oat, sweet vernal grass, Bermuda grass, rye grass, common reed, Kentucky blue grass, rye, wheat and maize (weak).
Phl p 1 binds to specific receptors on the surface of immune cells called TLR4 (Toll-like receptor 4) and MD-2 (myeloid differentiation factor 2). This binding triggers a signaling cascade that leads to the activation of immune cells and the release of inflammatory mediators, which cause the allergic symptoms. The interaction between Phl p 1 and TLR4-MD-2 complex has been studied extensively and is considered a key event in the development of grass pollen allergy. Understanding the molecular mechanisms involved in this interaction is important for developing new therapeutic strategies to treat or prevent allergic reactions.
Specifications
Specifications
| Antigen | Phl p 1 Chimeric |
| Applications | Dot Blot, ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | Clone 25 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene Alias | Group 1 grass pollen allergen; Phleum pratense allergen 1; Timothy grass pollen allergen |
| Host Species | Human |
| Immunogen | Recombinant Phl p 1, a grass pollen allergen. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?