missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-AMPK beta-1 (Ser182) Polyclonal Antibody
GREENER_CHOICE

Product Code. 11552410
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11552410 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11552410 Supplier Invitrogen™ Supplier No. PA517513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

AMPK beta 1 is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit may be a positive regulator of AMPK activity. The myristoylation and phosphorylation of this subunit have been shown to affect the enzyme activity and cellular localization of AMPK. This subunit may also serve as an adaptor molecule mediating the association of the AMPK complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-AMPK beta-1 (Ser182)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene PRKAB1
Gene Accession No. P80386, Q9R078, Q9Y478
Gene Alias 1300015D22Rik; 5'-AMP-activated protein kinase 40 kDa subunit; 5-AMP-activated protein kinase beta subunit; 5'-AMP-activated protein kinase beta-1 subunit; 5'-AMP-activated protein kinase subunit beta-1; 5'-AMP-activated protein kinase subunit beta-1-like protein; 5'-AMP-activated protein kinase, beta subunit; AAKB1; AMP-activated protein kinase beta subunit; AMPK; AMPK beta 1; AMPK beta -1 chain; AMPK beta-1 chain; AMPK subunit beta-1; AMPKb; AMPK-beta1; AU021155; E430008F22; HAMPKb; kinase AMPK-beta1; kinase pAMPK; pAMPK; Prkab1; protein kinase AMP-activated non-catalytic subunit beta 1; protein kinase, AMP-activated, beta 1 non-catalytic subunit; protein kinase, AMP-activated, noncatalytic, beta-1
Gene Symbols PRKAB1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pSer182 of human AMPKβ1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19079, 5564, 83803
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.