missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-B-Raf (Thr599) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17719963
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17719963 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17719963 Supplier Invitrogen™ Supplier No. PA5118542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of B Raf only when phosphorylated at Threonine 599.

BRAF is involved in the transduction of mitogenic signals from the cell membrane to the nucleus. It is activated by somatic point mutation in human cancer, and is thought to play a role in the postsynaptic responses of hippocampal neuron. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in B-Raf have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, malignant melanoma, thyroid carcinoma, non-small cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for B-Raf.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-B-Raf (Thr599)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BRAF
Gene Accession No. P15056, P28028
Gene Alias 94 kDa B-raf protein; 9930012E13Rik; AA120551; AA387315; AA473386; Braf; B-raf; B-raf protein; B-raf protein isoform 1; B-raf protein isoform 2; B-Raf proto-oncogene serine/threonine-protein kinase; B-Raf proto-oncogene serine/threonine-protein kinase (p94); B-Raf proto-oncogene, serine/threonine kinase; Braf transforming gene; BRAF1; B-RAF1; Braf2; Braf-2; C230098H17; C87398; C-RMIL; D6Ertd631e; FLJ95109; MGC126806; murine sarcoma viral (v-raf) oncogene homolog B1; NS7; p94; Proto-oncogene B-Raf; proto-oncogene c-Rmil; RAFB1; RMIL; rmil serine/threonine-protein kinase; serine/threonine kinase; serine/threonine-protein kinase B-raf; Serine/threonine-protein kinase Rmil; v-raf murine sarcoma viral oncogene homolog B; v-raf murine sarcoma viral oncogene homolog B1
Gene Symbols BRAF
Host Species Rabbit
Immunogen A synthesized peptide derived from human BRAF(Accession P15056), corresponding to amino acid residues around phosphorylated Thr599.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109880, 114486, 673
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.