missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-CaM Kinase II (T286) Antibody, R&D Systems™
Description
CaM Kinase II Polyclonal specifically detects CaM Kinase II in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | CaM Kinase II (p thr286) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 dilution |
| Host Species | Rabbit |
| Immunogen | Phosphopeptide corresponding to amino acid residues surrounding phospho-T286 of CaM Kinase II |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?