missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-MAPKAPK2 (Thr334) Recombinant Rabbit Monoclonal Antibody (MAPKAPK2T334-H2), APC, Invitrogen™

Product Code. 17156878 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
Unit Size:
Pack of 10
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17156878 10 Tests Pack of 10
17196598 100 Tests Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17156878 Supplier Invitrogen™ Supplier No. MA537289

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MAPKAPK2 mediates p38 and ERK signaling in vivo. It is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation, and cell proliferation. MAPKAPK2 encodes a member of the Ser/Thr protein kinase family. MAPKAPK2 is regulated through direct phosphorylation by p38 MAP kinase. In conjunction with p38 MAP kinase, MAPKAPK2 is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation and cell proliferation. Heat shock protein HSP27 was shown to be one of the substrates of this kinase in vivo. Two transcript variants encoding two different isoforms have been found for MAPKAPK2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MAPKAPK2 (Thr334)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone MAPKAPK2T334-H2
Conjugate APC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene MAPKAPK2
Gene Accession No. P49137, P49138
Gene Alias AA960234; MAP kinase-activated protein kinase 2; MAPK activated protein kinase 2; MAPK-activated protein kinase 2; MAPKAP kinase 2; Mapkapk2; MAPKAP-K2; MAPKAPK-2; mitogen-activated protein kinase-activated protein kinase 2; MK2; MK-2; Rps6kc1
Gene Symbols MAPKAPK2
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Thr334 of human phospho MAPKAPK-2.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17164, 9261
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.