missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MARCKS (Ser167/170) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29145-1-AP-20UL
This item is not returnable.
View return policy
Description
Phospho-MARCKS (Ser167/170) Polyclonal antibody specifically detects Phospho-MARCKS (Ser167/170) in Human samples. It is validated for WB, ELISASpecifications
| Phospho-MARCKS (Ser167/170) | |
| Polyclonal | |
| Unconjugated | |
| BC007688 | |
| Antigen affinity purification | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, ELISA | |
| 900 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Rabbit | |
| 20 μL | |
| 4082 | |
| Store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Condividi una correzione di contenuto