missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MARCKS (Ser167/170) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
135.00€ - 360.00€
Specifications
| Antigen | Phospho-MARCKS (Ser167/170) |
|---|---|
| Concentration | 900 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17781089
|
Proteintech
29145-1-AP-20UL |
20 μL |
135.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17771089
|
Proteintech
29145-1-AP-100UL |
100 μL |
360.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-MARCKS (Ser167/170) Polyclonal antibody specifically detects Phospho-MARCKS (Ser167/170) in Human samples. It is validated for WB, ELISASpecifications
| Phospho-MARCKS (Ser167/170) | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| IgG | |
| Antigen affinity purification | |
| Antibody |
| 900 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human | |
| 4082 | |
| Primary | |
| Store at -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title