missing translation for 'onlineSavingsMsg'
Learn More

Phospho-MLK3 (Ser674) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16642455
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16642455 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16642455 Supplier Invitrogen Supplier No. PA5106013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phospho-MLK3 (Ser674) Polyclonal Antibody

Phospho-MLK3 (Ser674) Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MLK3 (Ser674)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAP3K11
Gene Accession No. Q16584, Q66HA1, Q80XI6
Gene Alias 2610017K16Rik; MAP3K11; MEKK11; MGC17114; mitogen activated protein kinase kinase kinase 11; mitogen-activated protein kinase kinase kinase 11; Mixed lineage kinase 3; Mlk3; MLK-3; protein-tyrosine kinase PTK1; PTK1; RHOE; SH3 domain-containing proline-rich kinase; SPRK; Src-homology 3 domain-containing proline-rich kinase
Gene Symbols MAP3K11
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAP3K11(Accession Q16584), corresponding to amino acid residues around phosphorylated Ser674.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26403, 309168, 4296
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.