missing translation for 'onlineSavingsMsg'
Learn More

Phospho-TBK1 (Ser172) Rabbit Recombinant Antibody, Proteintech

Product Code. p-200107070 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235036 20 μL 20µL
30238088 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235036 Supplier Proteintech Supplier No. 823831RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Rabbit Recombinant Antibody

The Phospho-TBK1 (Ser172) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-TBK1 (Ser172). This antibody recognizes human antigen. The Phospho-TBK1 (Ser172) antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen TBK1 (Ser172)
Applications Western Blot, ELISA
Classification Recombinant Monoclonal
Clone 2K2
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols FLJ11330; NAK; T2K
Host Species Rabbit
Immunogen Peptide
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29110
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 84 kDa
Product Type Recombinant Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.